APO455826203 Apollo Scientific Ltd

4-Aminobenzeneboronic acid, N-CBZ protected

  • MDL number MFCD02093055
  • Cas Number : 192804-36-7
  • Molecular Weight : 271.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 168-169
Iupac Name 4-{[(benzyloxy)carbonyl]amino}phenylboronic acid
Inchi Code 1S/C14H14BNO4/c17-14(20-10-11-4-2-1-3-5-11)16-13-8-6-12(7-9-13)15(18)19/h1-9,18-19H,10H2,(H,16,17)
Inchi Key KYCQBMOJDXVNIC-UHFFFAOYSA-N

Description

Synonym 4-{[(Benzyloxy)carbonyl]amino}benzeneboronic acid, Benzyl N-(4-boronophenyl)carbamate
Purity 96%
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 168-169
Product Link https://store.apolloscientific.co.uk/product/4-aminobenzeneboronic-acid-n-cbz-protected

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-aminobenzeneboronic-acid-n-cbz-protected
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart