APO455826207 Apollo Scientific Ltd

5-Chloro-2-methylbenzeneboronic acid

  • MDL number MFCD03411939
  • Cas Number : 148839-33-2
  • Molecular Weight : 170.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 184-186
Purity 97
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-chloro-2-methylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-chloro-2-methylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart