APO455826212 Apollo Scientific Ltd

4-(Cyclohexylcarbamoyl)benzeneboronic acid

  • MDL number MFCD03411956
  • Cas Number : 762262-07-7
  • Molecular Weight : 247.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 174-178
Synonym 4-[(Cyclohexylamino)carbonyl]benzeneboronic acid, 4-Borono-N-cyclohexylbenzamide
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-cyclohexylcarbamoylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-cyclohexylcarbamoylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart