APO455826214 Apollo Scientific Ltd

1-Methyl-1H-imidazole-4,5-dicarboxylic acid

  • MDL number MFCD01552045
  • Cas Number : 19485-38-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4,5-Dicarboxy-1-methyl-1H-imidazole
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=16592

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=16592
Availability details for
Quantity
  • Availability
  • Add to Cart