APO455826220 Apollo Scientific Ltd

4-(Diethylcarbamoyl)benzeneboronic acid

  • MDL number MFCD03411949
  • Cas Number : 389621-80-1
  • Molecular Weight : 221.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138-142
Synonym 4-Borono-N,N-diethylbenzamide, 4-[(Diethylamino)carbonyl]benzeneboronic acid
Purity 98
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-diethylcarbamoylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-diethylcarbamoylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart