APO455826221 Apollo Scientific Ltd

4,5-Diiodo-1H-imidazole

  • MDL number MFCD00963878
  • Cas Number : 15813-09-9
  • Molecular Weight : 319.87

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180-181
Purity 98
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/45-diiodo-1h-imidazole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/45-diiodo-1h-imidazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart