APO455826223 Apollo Scientific Ltd

3-(Dimethylcarbamoyl)benzeneboronic acid

  • MDL number MFCD03412066
  • Cas Number : 373384-14-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 122-128
Synonym 3-(Dimethylaminocarbonyl)benzeneboronic acid, N,N-Dimethyl 3-boronobenzamide
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=16608

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=16608
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart