APO455826225 Apollo Scientific Ltd

2,3-Dimethylbenzeneboronic acid

  • MDL number MFCD01863524
  • Cas Number : 183158-34-1
  • Molecular Weight : 149.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 174-180
Iupac Name 2,3-dimethylphenylboronic acid
Inchi Code 1S/C8H11BO2/c1-6-4-3-5-8(7(6)2)9(10)11/h3-5,10-11H,1-2H3
Inchi Key ZYYANAWVBDFAHY-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 174-180
Product Link https://store.apolloscientific.co.uk/product/23-dimethylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P280;P302+P352;P304+P340;P305+P351+P338;P312;P321
MSDS https://store.apolloscientific.co.uk/product/23-dimethylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart