APO455826239 Apollo Scientific Ltd

Ethyl 3-amino-1H-pyrrole-2-carboxylate hydrochloride

  • MDL number MFCD04038917
  • Cas Number : 252932-49-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 198-204
Synonym 3-Amino-2-(ethoxycarbonyl)-1H-pyrrole hydrochloride
Purity 95
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=16642

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=16642
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart