APO455826242 Apollo Scientific Ltd

4-Hydroxy-2-methylbenzeneboronic acid

  • MDL number MFCD03788424
  • Cas Number : 493035-82-8
  • Molecular Weight : 151.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100-104
Synonym 4-Borono-3-methylphenol, 2-Borono-5-hydroxytoluene
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-hydroxy-2-methylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-hydroxy-2-methylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart