APO455826254 Apollo Scientific Ltd

4-Iodo-1-methyl-1H-imidazole

  • MDL number MFCD01632214
  • Cas Number : 71759-87-0
  • Molecular Weight : 208

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 51-54
Purity 97
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-iodo-1-methyl-1h-imidazole

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-iodo-1-methyl-1h-imidazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart