APO455826259 Apollo Scientific Ltd

2-Isopropylbenzeneboronic acid

  • MDL number MFCD03411937
  • Cas Number : 89787-12-2
  • Molecular Weight : 164.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 74-78
Synonym 2-(Prop-2-yl)benzeneboronic acid, o-Cumylboronic acid
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-isopropylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-isopropylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart