APO455826260 Apollo Scientific Ltd

3-Isopropylbenzeneboronic acid

  • MDL number MFCD01074634
  • Cas Number : 216019-28-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 72-76
Synonym (3-Isopropylphenyl)boronic acid
Purity 98
Flash Point >110
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=16674

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=16674
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart