APO455826274 Apollo Scientific Ltd

Methyl 1-methyl-1H-imidazole-4-carboxylate

  • MDL number MFCD02179561
  • Cas Number : 17289-19-9
  • Molecular Weight : 140.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 101.5-103.5
Synonym 4-(Methoxycarbonyl)-1-methyl-1H-imidazole
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-1-methyl-1h-imidazole-4-carboxylate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-1-methyl-1h-imidazole-4-carboxylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart