APO455826276 Apollo Scientific Ltd

Methyl oxindole-7-carboxylate

  • MDL number MFCD02179610
  • Cas Number : 380427-39-4
  • Molecular Weight : 191.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 182-184
Purity 96
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-oxindole-7-carboxylate

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-oxindole-7-carboxylate
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart