APO455826278 Apollo Scientific Ltd

3-(Methylcarbamoyl)benzeneboronic acid

  • MDL number MFCD04038918
  • Cas Number : 832695-88-2
  • Molecular Weight : 178.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 114-124
Synonym 3-Borono-N-methylbenzamide
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-methylcarbamoylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-methylcarbamoylbenzeneboronic-acid
pictograms GHS07
hstatements H302;H312;H332
Availability details for
Quantity
  • Availability
  • Add to Cart