APO455826279 Apollo Scientific Ltd

4-(Methylcarbamoyl)benzeneboronic acid

  • MDL number MFCD03788427
  • Cas Number : 121177-82-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 200-220
Synonym 4-Borono-N-methylbenzamide, 4-[(Methylamino)carbonyl]benzeneboronic acid
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-methylcarbamoylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-methylcarbamoylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart