APO455826281 Apollo Scientific Ltd

6-(Methylsulphonyl)-2-oxindole

  • MDL number MFCD02179607
  • Cas Number : 850429-63-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 218-220
Synonym 1,3-Dihydro-6-(methylsulphonyl)-2H-indol-2-one, 6-(Methylsulphonyl)indolin-2-one
Purity 97
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=16707

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=16707
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart