APO455826287 Apollo Scientific Ltd

2-(Methylsulphonyl)benzeneboronic acid

  • MDL number MFCD02179472
  • Cas Number : 330804-03-0
  • Molecular Weight : 200.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 78-84
Iupac Name 2-(methylsulfonyl)phenylboronic acid
Inchi Code 1S/C7H9BO4S/c1-13(11,12)7-5-3-2-4-6(7)8(9)10/h2-5,9-10H,1H3
Inchi Key MSQFFCRGQPVQRS-UHFFFAOYSA-N

Description

Synonym 2-Boronophenyl methyl sulphone
Purity 97%
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 78-84
Product Link https://store.apolloscientific.co.uk/product/2-methylsulphonylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-methylsulphonylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart