APO455826291 Apollo Scientific Ltd

4-[(Morpholin-4-yl)carbonyl]benzeneboronic acid

  • MDL number MFCD03411952
  • Cas Number : 389621-84-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 126-132
Synonym 4-(4-Boronobenzoyl)morpholine, (4-Boronophenyl)(morpholin-4-yl)methanone
Purity 98
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=16718

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=16718
pictograms GHS07
hstatements H315;H319;H335;H302
Availability details for
Quantity
  • Availability
  • Add to Cart