APO455826300 Apollo Scientific Ltd

4-[(Pyrrolidin-1-yl)carbonyl]benzeneboronic acid

  • MDL number MFCD03411950
  • Cas Number : 389621-81-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 136-140
Synonym (4-Boronophenyl)(pyrrolidin-1-yl)methanone, 1-(4-Boronobenzoyl)pyrrolidine
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=16734

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=16734
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart