APO455826301 Apollo Scientific Ltd

4-(Hydroxymethyl)tetrahydro-2H-pyran

  • MDL number MFCD00457804
  • Cas Number : 14774-37-9
  • Molecular Weight : 116.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (Tetrahydro-2H-pyran-4-yl)methanol, (Oxan-4-yl)methanol
Purity 98
Flash Point >61
Physical Form Liquid-Clear
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 105-110/20mm
Product Link https://store.apolloscientific.co.uk/product/4-hydroxymethyltetrahydro-2h-pyran

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-hydroxymethyltetrahydro-2h-pyran
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart