APO455826306 Apollo Scientific Ltd

1-Trityl-1H-imidazole-4-carboxaldehyde

  • MDL number MFCD02179554
  • Cas Number : 33016-47-6
  • Molecular Weight : 338.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 187-192(dec.)
Synonym 4-Formyl-1-trityl-1H-imidazole, (4-Formyl-1H-imidazol-1-yl)triphenylmethane
Purity 95
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-trityl-1h-imidazole-4-carboxaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/1-trityl-1h-imidazole-4-carboxaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart