APO455826308 Apollo Scientific Ltd

2-Vinylbenzeneboronic acid

  • MDL number MFCD01074659
  • Cas Number : 15016-42-9
  • Molecular Weight : 147.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105-107
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-vinylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P280;P261
MSDS https://store.apolloscientific.co.uk/product/2-vinylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart