APO455826323 Apollo Scientific Ltd

2-Amino-4-chloro-6-isopropylpyrimidine

  • MDL number MFCD03840198
  • Cas Number : 73576-33-7
  • Molecular Weight : 171.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 97-101
Synonym 4-Chloro-6-(prop-2-yl)pyrimidin-2-amine, 4-Chloro-6-isopropylpyrimidin-2-amine
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-amino-4-chloro-6-isopropylpyrimidine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-4-chloro-6-isopropylpyrimidine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart