APO455826330 Apollo Scientific Ltd

(3-Ethoxy-3-oxopropyl)(triphenyl)phosphonium bromide

  • MDL number MFCD03701355
  • Cas Number : 42843-94-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 145(dec.)
Purity 97
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-ethoxy-3-oxopropyltriphenylphosphonium-bromide

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-ethoxy-3-oxopropyltriphenylphosphonium-bromide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart