APO455826331 Apollo Scientific Ltd

2,6-Dimethyl-4-hydroxypyridine

  • MDL number MFCD00956021
  • Cas Number : 13603-44-6
  • Molecular Weight : 123.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 230
Synonym 2,6-Dimethylpyridin-4-ol, 4-Hydroxy-2,6-lutidine
Purity 98
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 351
Product Link https://store.apolloscientific.co.uk/product/26-dimethyl-4-hydroxypyridine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/26-dimethyl-4-hydroxypyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart