APO455826336 Apollo Scientific Ltd

(Methoxycarbonylmethyl)tris(phenyl)phosphonium chloride

  • MDL number MFCD00050199
  • Cas Number : 2181-97-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 155(dec.)
Synonym (2-Methoxy-2-oxoethyl)tris(phenyl)phosphonium chloride
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=16811

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=16811
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart