APO455826337 Apollo Scientific Ltd

(Methoxymethyl)(triphenyl)phosphonium bromide

  • MDL number MFCD00075442
  • Cas Number : 33670-32-5
  • Molecular Weight : 387.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 230
Iupac Name (methoxymethyl)(triphenyl)phosphonium bromide
Inchi Code 1S/C20H20OP.BrH/c1-21-17-22(18-11-5-2-6-12-18,19-13-7-3-8-14-19)20-15-9-4-10-16-20;/h2-16H,17H2,1H3;1H/q+1;/p-1
Inchi Key NOCGROPYCGRERZ-UHFFFAOYSA-M

Description

Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 230
Product Link https://store.apolloscientific.co.uk/product/methoxymethyltriphenylphosphonium-bromide

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methoxymethyltriphenylphosphonium-bromide
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart