APO455826347 Apollo Scientific Ltd

Diethyl (but-1-yl)phosphonate

  • MDL number MFCD08445506
  • Cas Number : 2404-75-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 74-76/1mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=16844

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=16844
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart