APO455826351 Apollo Scientific Ltd

Diethyl dodec-1-ylphosphonate

  • MDL number MFCD00015595
  • Cas Number : 4844-38-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 168/1.5mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=16856

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=16856
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart