APO455826373 Apollo Scientific Ltd

3-Methylbenzylphosphonic acid

  • MDL number MFCD03411186
  • Cas Number : 18945-65-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 194-195
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=16933

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=16933
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart