APO455826376 Apollo Scientific Ltd

4-Chloro-3-methylbenzoic acid

  • MDL number MFCD00045853
  • Cas Number : 7697-29-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 212-216
Synonym 4-Chloro-m-toluic acid, 5-Carboxy-2-chlorotoluene
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-chloro-3-methylbenzoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-chloro-3-methylbenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart