APO455826379 Apollo Scientific Ltd

Dimethyl hydrogen phosphate

  • MDL number MFCD00014887
  • Cas Number : 813-78-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Dimethyl phosphate
Density 1.323
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 174
Product Link http://www.apolloscientific.co.uk/display_item.php?id=16954

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=16954
pictograms GHS05;GHS07
hstatements H312;H314
Availability details for
Quantity
  • Availability
  • Add to Cart