APO455826388 Apollo Scientific Ltd

4-Bromophenoxyacetaldehyde diethylacetal

  • MDL number MFCD03411183
  • Cas Number : 112598-18-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Bromo-4-(2,2-diethoxyethoxy)benzene
Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=16981

Safety Information

signalword Warning
pcodes P280;P312
MSDS http://www.apolloscientific.co.uk/display_item.php?id=16981
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart