APO455826389 Apollo Scientific Ltd

3-Bromo-2-oxopropanoic acid hydrate

  • MDL number MFCD00149107
  • Cas Number : 206860-50-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 79-82
Synonym 3-Bromopyruvic acid hydrate
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=16984

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=16984
pictograms GHS05;GHS07
hstatements H302+H312+H332;H314;H318
Availability details for
Quantity
  • Availability
  • Add to Cart