APO455826402 Apollo Scientific Ltd

3-[(Dimethylcarbamoyl)amino]benzoic acid

  • MDL number MFCD03411196
  • Cas Number : 896160-52-4
  • Molecular Weight : 208.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-(3,3-Dimethylureido)benzoic acid, 3-(3-Carboxyphenyl)-1,1-dimethylurea
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-dimethylcarbamoylaminobenzoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-dimethylcarbamoylaminobenzoic-acid
pictograms GHS07;GHS05
hstatements H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart