APO455826408 Apollo Scientific Ltd

Benzo[b]furan-5-carboxaldehyde

  • MDL number MFCD03411182
  • Cas Number : 10035-16-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 36-38
Synonym 5-Formyl-1-benzofuran, 5-Formylbenzo[b]furan, 1-Benzofuran-5-carboxaldehyde
Purity 97
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=17023

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=17023
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart