APO455826411 Apollo Scientific Ltd

4-[(Methylsulphonyl)amino]benzoic acid

  • MDL number MFCD00025052
  • Cas Number : 7151-76-0
  • Molecular Weight : 215.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-(Methylsulphonamido)benzoic acid, N-(4-Carboxyphenyl)methanesulphonamide
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-methylsulphonylaminobenzoic-acid

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-methylsulphonylaminobenzoic-acid
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart