APO455826427 Apollo Scientific Ltd

4-Bromo-3-nitrobenzeneboronic acid

  • MDL number MFCD01074649
  • Cas Number : 74386-13-3
  • Molecular Weight : 245.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 213-216
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-3-nitrobenzeneboronic-acid

Safety Information

signalword Warning
pcodes P260;P280;P312
MSDS https://store.apolloscientific.co.uk/product/4-bromo-3-nitrobenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart