APO455826431 Apollo Scientific Ltd

Benzyl ethyl piperidine-1,3-dicarboxylate

  • MDL number MFCD06657752
  • Cas Number : 310454-53-6
  • Molecular Weight : 291.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/benzyl-ethyl-piperidine-13-dicarboxylate

Safety Information

signalword Warning
pcodes P261;P280;P271
MSDS https://store.apolloscientific.co.uk/product/benzyl-ethyl-piperidine-13-dicarboxylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart