APO455826444 Apollo Scientific Ltd

Decane-1,10-diamine, N-BOC protected

  • MDL number MFCD02094501
  • Cas Number : 216961-61-4
  • Molecular Weight : 272.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 10-aminodecylcarbamate
Inchi Code 1S/C15H32N2O2/c1-15(2,3)19-14(18)17-13-11-9-7-5-4-6-8-10-12-16/h4-13,16H2,1-3H3,(H,17,18)
Inchi Key RSAPJHYIFKJREV-UHFFFAOYSA-N

Description

Synonym tert-Butyl (10-aminodec-1-yl)carbamate, 1,10-Diaminodecane, N-BOC protected
Purity 97%
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/decane-110-diamine-n-boc-protected

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/decane-110-diamine-n-boc-protected
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart