APO455826446 Apollo Scientific Ltd

3-Phenyl-2-thioxo-1,3-thiazolidin-4-one

  • MDL number MFCD00100196
  • Cas Number : 1457-46-1
  • Molecular Weight : 209.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 190-194
Synonym N-Phenyl-rhodanine
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-phenyl-2-thioxo-13-thiazolidin-4-one

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-phenyl-2-thioxo-13-thiazolidin-4-one
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart