APO455826456 Apollo Scientific Ltd

3-Amino-3-(4-hydroxyphenyl)propanoic acid

  • MDL number MFCD00181810
  • Cas Number : 6049-54-3
  • Molecular Weight : 181.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 193-195(dec.)
Synonym 3-Amino-3-(4-hydroxyphenyl)propionic acid, 4-(1-Amino-2-carboxyethyl)phenol, beta-Amino-4-hydroxyhydrocinnamic acid
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-amino-3-4-hydroxyphenylpropanoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-amino-3-4-hydroxyphenylpropanoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart