APO455826458 Apollo Scientific Ltd

Heptane-1,7-diamine, N-BOC protected

  • MDL number MFCD02094498
  • Cas Number : 99733-18-3
  • Molecular Weight : 230.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 113-114
Iupac Name tert-butyl 7-aminoheptylcarbamate
Inchi Code 1S/C12H26N2O2/c1-12(2,3)16-11(15)14-10-8-6-4-5-7-9-13/h4-10,13H2,1-3H3,(H,14,15)
Inchi Key DTJYPERGUPPXRU-UHFFFAOYSA-N

Description

Synonym tert-Butyl (7-aminohept-1-yl)carbamate, 1,7-Diaminoheptane, N-BOC protected
Purity 95%
Flash Point 96-100/0.1mm
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 113-114
Product Link https://store.apolloscientific.co.uk/product/heptane-17-diamine-n-boc-protected

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/heptane-17-diamine-n-boc-protected
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart