APO455826467 Apollo Scientific Ltd

4-Methylbenzeneboronic acid, pinacol ester

  • MDL number MFCD03453662
  • Cas Number : 195062-57-8
  • Molecular Weight : 218.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 54
Synonym 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)toluene, 2-(4-Methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-methylbenzeneboronic-acid-pinacol-ester

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-methylbenzeneboronic-acid-pinacol-ester
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart