APO455826472 Apollo Scientific Ltd

3,5-Dimethyl-4-iodophenol

  • MDL number MFCD00174284
  • Cas Number : 80826-86-4
  • Molecular Weight : 248.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 129-130
Synonym 5-Hydroxy-2-iodo-m-xylene, 4-Iodo-3,5-xylenol
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/35-dimethyl-4-iodophenol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/35-dimethyl-4-iodophenol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart