APO455826481 Apollo Scientific Ltd

4'-Chlorovalerophenone

  • MDL number MFCD00018716
  • Cas Number : 25017-08-7
  • Molecular Weight : 196.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 29-33
Iupac Name 1-(4-chlorophenyl)-1-pentanone
Inchi Code 1S/C11H13ClO/c1-2-3-4-11(13)9-5-7-10(12)8-6-9/h5-8H,2-4H2,1H3
Inchi Key XMUGWCSIQUJOFA-UHFFFAOYSA-N

Description

Synonym 1-(4-Chlorophenyl)pentan-1-one
Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 29-33
Product Link https://store.apolloscientific.co.uk/product/4-chlorovalerophenone

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS https://store.apolloscientific.co.uk/product/4-chlorovalerophenone
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart