APO455826490 Apollo Scientific Ltd

2-Chlorophenoxyacetic acid hydrazide

  • MDL number MFCD00082786
  • Cas Number : 36304-40-2
  • Molecular Weight : 200.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 111-112
Purity 98
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-chlorophenoxyacetic-acid-hydrazide

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-chlorophenoxyacetic-acid-hydrazide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart