APO455826492 Apollo Scientific Ltd

Ethyl 2-amino-5-iodobenzoate

  • MDL number MFCD04038940
  • Cas Number : 268568-11-2
  • Molecular Weight : 291.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 70-73
Synonym Ethyl 5-iodoanthranilate, 2-(Ethoxycarbonyl)-4-iodoaniline
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-2-amino-5-iodobenzoate

Safety Information

signalword Warning
pcodes P280;P304+P340
MSDS https://store.apolloscientific.co.uk/product/ethyl-2-amino-5-iodobenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart